AC5414275632 Accela ChemBio Inc.

5-Methoxy-2-thiophenecarboxylic Acid

  • MDL number MFCD02317367
  • Cas Number : 29212-22-4
  • Molecular Weight : 158.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022425&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022425.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart