AC5414275639 Accela ChemBio Inc.

Benzyl 5-[Methoxy(methyl)amino]-5-oxopentanoate

  • MDL number MFCD22683326
  • Cas Number : 1418117-77-7
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022563&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022563.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart