AC5414275640 Accela ChemBio Inc.

Succinic Acid Monobenzyl Ester

  • MDL number MFCD00447717
  • Cas Number : 103-40-2
  • Molecular Weight : 208.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022564&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022564.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart