AC5414275648 Accela ChemBio Inc.

Ethyl 5-Bromo-4-pyrimidinecarboxylate

  • MDL number MFCD16660482
  • Cas Number : 64224-59-5
  • Molecular Weight : 231.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022729&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022729.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart