AC5414275661 Accela ChemBio Inc.

4-Bromo-2-(trifluoromethyl)benzoyl Chloride

  • MDL number MFCD13185394
  • Cas Number : 104356-17-4
  • Molecular Weight : 287.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023645&attname=GoodsCode

Safety Information

signalword Danger
hcodes GHS05, GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY023645.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart