AC5414275662 Accela ChemBio Inc.

1,5-Dimethyl-4-nitroimidazole

  • MDL number MFCD01721572
  • Cas Number : 7464-68-8
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023762&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023762.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart