AC5414275671 Accela ChemBio Inc.

Benzofuran-7-carbaldehyde

  • MDL number MFCD08272125
  • Cas Number : 95333-14-5
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024200&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P302+P352-P280-P305+P351+P338
MSDS http://pdf.accelachem.com/cn/msds/SY024200.pdf
pictograms GHS07
hstatements H315-H319-H335-H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart