AC5414275692 Accela ChemBio Inc.

1,2-Phenylenediacetonitrile

  • MDL number MFCD00001905
  • Cas Number : 613-73-0
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025677&attname=GoodsCode

Safety Information

signalword Danger
pcodes P280H-P309-P310
hcodes GHS06
MSDS http://pdf.accelachem.com/cn/msds/SY025677.pdf
hstatements H301-H311-H331
Availability details for
Quantity
  • Availability
  • Add to Cart