AC5414275697 Accela ChemBio Inc.

8-Bromoisoquinoline Hydrochloride

  • MDL number MFCD20486979
  • Cas Number : 1307316-93-3
  • Molecular Weight : 244.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025847&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025847.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart