AC5414275715 Accela ChemBio Inc.

7-Chloro-1H-pyrido[3,4-b][1,4]oxazin-2(3H)-one

  • MDL number MFCD11044713
  • Cas Number : 928118-43-8
  • Molecular Weight : 184.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025992&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025992.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart