AC5414275716 Accela ChemBio Inc.

(S)-2-Methylpyrrolidine-2-carboxamide

  • MDL number MFCD19204326
  • Cas Number : 132235-43-9
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025997&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025997.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart