AC5414275722 Accela ChemBio Inc.

4-Amino-6-chloro-2-methylpyrimidine-5-carbonitrile

  • MDL number MFCD07761831
  • Cas Number : 76574-37-3
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026327&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026327.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart