AC5414275724 Accela ChemBio Inc.

Methyl 3-Chloro-5-formylbenzoate

  • MDL number MFCD22544335
  • Cas Number : 879542-48-0
  • Molecular Weight : 198.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026331&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026331.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart