AC5414275726 Accela ChemBio Inc.

3-Fluoro-5-formylbenzoic Acid

  • MDL number MFCD22544368
  • Cas Number : 1289005-85-1
  • Molecular Weight : 168.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026335&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026335.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart