AC5414275729 Accela ChemBio Inc.

4-Aminopyridine-2-carboxylic Acid Hydrochloride Hydrate

  • MDL number MFCD26523293
  • Molecular Weight : 192.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026378&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026378.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart