AC5414275733 Accela ChemBio Inc.

3-(Fluoromethyl)aniline

  • MDL number MFCD00466952
  • Cas Number : 456-46-2
  • Molecular Weight : 125.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026432&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY026432.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart