AC5414275735 Accela ChemBio Inc.

2-Amino-3-hydroxy-3-methylbutanoic Acid

  • MDL number MFCD00145237
  • Cas Number : 2280-28-6
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026436&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026436.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart