AC5414275742 Accela ChemBio Inc.

5-Chloro-4-methoxysalicylaldehyde

  • MDL number MFCD08059080
  • Cas Number : 89938-56-7
  • Molecular Weight : 186.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026584&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026584.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart