AC5414275743 Accela ChemBio Inc.

2-Amino-1-Boc-imidazole

  • MDL number MFCD20702858
  • Cas Number : 929568-19-4
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026587&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026587.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart