AC5414275762 Accela ChemBio Inc.

5,6-Dichloro-2-(3-hydroxypropyl)benzimidazole

  • MDL number MFCD11106864
  • Cas Number : 6478-83-7
  • Molecular Weight : 245.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026778&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026778.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart