AC5414275765 Accela ChemBio Inc.

2-Amino-2-methylpropanamide

  • MDL number MFCD11505290
  • Cas Number : 16252-90-7
  • Molecular Weight : 102.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026852&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026852.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart