AC5414275792 Accela ChemBio Inc.

Methyl 3-[(4-Hydroxyphenyl)amino]propanoate

  • MDL number MFCD20666958
  • Cas Number : 70156-40-0
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow or pink solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027115&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027115.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart