AC5414275793 Accela ChemBio Inc.

1-[3-(Dimethylamino)propyl]-3-ethylurea

  • MDL number MFCD00027752
  • Cas Number : 32897-26-0
  • Molecular Weight : 173.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027122&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280a-P305+P351+P338-P304+P340-P405-P501a
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY027122.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart