AC5414275794 Accela ChemBio Inc.

3-(Aminomethyl)-2-naphthol

  • MDL number MFCD18410094
  • Cas Number : 1181563-65-4
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027128&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027128.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart