AC5414275795 Accela ChemBio Inc.

5-Bromo-3-methylbenzofuran

  • MDL number MFCD09836519
  • Cas Number : 33118-85-3
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027158&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027158.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart