AC5414275796 Accela ChemBio Inc.

5-(2,6-Dimethylphenyl)indoline

  • MDL number MFCD11878807
  • Cas Number : 1176740-73-0
  • Molecular Weight : 223.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027164&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027164.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart