AC5414275806 Accela ChemBio Inc.

2-(5-Hydroxybenzo[d]isoxazol-3-yl)acetic Acid

  • MDL number MFCD20639829
  • Cas Number : 34173-03-0
  • Molecular Weight : 193.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027287&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027287.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart