AC5414275814 Accela ChemBio Inc.

1,8-Naphthalenediyl Dibenzoate

  • MDL number MFCD01001009
  • Cas Number : 331711-99-0
  • Molecular Weight : 368.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027344&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027344.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart