AC5414275821 Accela ChemBio Inc.

Methyl 2-Phenylpiperidine-4-carboxylate

  • MDL number MFCD07367319
  • Cas Number : 351003-06-0
  • Molecular Weight : 219.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027394&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027394.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart