AC5414275822 Accela ChemBio Inc.

Ethyl N-Boc-2,5-dihydropyrrole-3-carboxylate

  • MDL number MFCD11973978
  • Cas Number : 146257-00-3
  • Molecular Weight : 241.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027395&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027395.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart