AC5414275824 Accela ChemBio Inc.

Ethyl 3-(3-Hydroxyphenyl)-3-oxopropanoate

  • MDL number MFCD24718690
  • Cas Number : 77103-49-2
  • Molecular Weight : 208.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027397&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027397.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart