AC5414275825 Accela ChemBio Inc.

Methyl N-Boc-2-amino-6-bromohexanoate

  • MDL number MFCD24472605
  • Molecular Weight : 324.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027403&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027403.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart