AC5414275832 Accela ChemBio Inc.

2-[4-(2-Pyrimidyl)-1-piperazinyl]acetohydrazide

  • MDL number MFCD19386499
  • Cas Number : 1495336-65-6
  • Molecular Weight : 236.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027504&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027504.pdf
pictograms No data known
Availability details for
Quantity
  • Availability
  • Add to Cart