AC5414275838 Accela ChemBio Inc.

2-Amino-4-iodobenzoic Acid

  • MDL number MFCD09800365
  • Cas Number : 20776-54-9
  • Molecular Weight : 263.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027629&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027629.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart