AC5414275841 Accela ChemBio Inc.

N-Boc-1-methylcyclopropanamine

  • MDL number MFCD11111233
  • Cas Number : 251661-01-5
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027633&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027633.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart