AC5414275848 Accela ChemBio Inc.

1-(3-Fluorophenyl)cyclohexanol

  • MDL number MFCD00218768
  • Cas Number : 1496-35-1
  • Molecular Weight : 194.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027682&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027682.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart