AC5414275860 Accela ChemBio Inc.

1-Cbz-2-methylpyrrolidine-2-carboxylic Acid

  • MDL number MFCD11215478
  • Cas Number : 63427-91-8
  • Molecular Weight : 263.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027954&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027954.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart