AC5414275861 Accela ChemBio Inc.

Ethyl 3-(4-Hydroxyphenyl)-5-methylisoxazole-4-carboxylate

  • MDL number MFCD22372653
  • Cas Number : 1071788-87-8
  • Molecular Weight : 247.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027963&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027963.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart