AC5414275864 Accela ChemBio Inc.

Methyl 2-[4-(-2-Pyrimidyl)-1-piperazinyl]acetate

  • MDL number MFCD05071221
  • Cas Number : 896213-29-9
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027979&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027979.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart