AC5414275866 Accela ChemBio Inc.

Ethyl 3-Amino-3-(3-hydroxyphenyl)propanoate

  • MDL number MFCD02110516
  • Cas Number : 309735-43-1
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028064&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028064.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart