AC5414275869 Accela ChemBio Inc.

2-Phenoxyacetamidine Hydrochloride

  • MDL number MFCD00173806
  • Cas Number : 67386-38-3
  • Molecular Weight : 186.63874

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003970&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P302+P352-P261-P301+P312-P304+P340-P280-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY003970.pdf
hstatements H302-H312-H315-H332-H335-H319
Availability details for
Quantity
  • Availability
  • Add to Cart