AC5424073115 Accela ChemBio Inc.

3-Piperidylacetic Acid Hydrochloride

  • MDL number MFCD02181158
  • Cas Number : 71985-81-4
  • Molecular Weight : 179.64456

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY002543&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY002543.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart