AC5424073164 Accela ChemBio Inc.

cis-1-Methylhexahydropyrrolo[3,4-b]pyrrole Dihydrochloride

  • MDL number MFCD21364418
  • Cas Number : 1234805-59-4
  • Molecular Weight : 199.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003868&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY003868.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart