AC5424073166 Accela ChemBio Inc.

(3aR,6aR)-1-Methyl-hexahydropyrrolo[3,4-b]pyrrole Dihydrochloride

  • MDL number MFCD11656764
  • Cas Number : 1417789-76-4
  • Molecular Weight : 199.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003871&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY003871.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart