AC5424073188 Accela ChemBio Inc.

(S)-2-(1-Pyrrolidyl)propanoic Acid

  • MDL number MFCD11656775
  • Cas Number : 786627-71-2
  • Molecular Weight : 143.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004138&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004138.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart