AC5424073237 Accela ChemBio Inc.

(S)-4-Benzyl-3-morpholinecarboxylic Acid

  • MDL number MFCD06799463
  • Cas Number : 1235011-96-7
  • Molecular Weight : 221.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005133&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY005133.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart