AC5424073252 Accela ChemBio Inc.

3,4-Dihydro-2H-1,5-benzodioxepin-7-amine

  • MDL number MFCD00218806
  • Cas Number : 175136-34-2
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005333&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P301+P310-P305+P351+P338-P361-P405-P501A
hcodes GHS07, GHS09
MSDS http://pdf.accelachem.com/cn/msds/SY005333.pdf
hstatements H301-H311-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart