AC5424073271 Accela ChemBio Inc.

3-Amino-5-fluorobenzo[d]isoxazole

  • MDL number MFCD09999137
  • Cas Number : 868271-13-0
  • Molecular Weight : 152.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005534&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY005534.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart