AC5424073285 Accela ChemBio Inc.

Methyl 4-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate

  • MDL number MFCD12198873
  • Cas Number : 196301-95-8
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005718&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY005718.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart