AC5424073291 Accela ChemBio Inc.

Methyl 4-(5-Isopropyl-1,2,4-oxadiazol-3-yl)benzoate

  • MDL number MFCD12198878
  • Cas Number : 1166756-82-6
  • Molecular Weight : 246.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005727&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY005727.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart